missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ UK 383367
GREENER_CHOICE

Product Code. 18095322
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18095322 1 mg 1mg
18034462 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18095322 Supplier Tocris Bioscience™ Supplier No. 4188/1
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective BMP-1 (PCP) inhibitor

Potent and selective inhibitor of BMP-1 (procollagen C-proteinase; PCP) (IC50 = 44 nM). Selective for BMP-1 over MMPs 1, 2, 3, 9 and 14 (IC50 values are >10,000 nM for listed MMPs).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348622-88-8
Molecular Formula C15H24N4O4

Specifications

Quantity 1 mg
Formula Weight 324.38
Purity >99%
Molecular Weight (g/mol) 324.381
Chemical Name or Material UK 383367
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.