missing translation for 'onlineSavingsMsg'
Learn More

UK-371804, MedChemExpress

Product Code. 16493298
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493298 10 mg 10mg
16489836 50 mg 50mg
16416027 5 mg 5mg
16404556 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493298 Supplier Medchem Express Supplier No. HY101214
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress UK-371804 is a urokinase-type plasminogen activator (uPA) inhibitor with a Ki of 10 nM.

UK-371804 is a urokinase-type plasminogen activator (uPA) inhibitor with a Ki of 10 nM.

Specifications

Chemical Name or Material UK-371804
CAS 256477-09-5
Color White
Purity Grade Notes Research
Molecular Formula C14H16ClN5O4S
Quantity 10 mg
Solubility Information DMSO : 8.33 mg/mL (21.59 mM; Need ultrasonic)
SMILES CC(C(O)=O)(C)NS(=O)(C1=CC2=C(C=C1)C(Cl)=CN=C2NC(N)=N)=O
Molecular Weight (g/mol) 385.83
Formula Weight 385.83
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.