missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UHRF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580206
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat testis tissue, K562 whole cell. IHC: mouse intestine tissue, rat intestine tissue, human intestinal cancer tissue.
This gene encodes a nuclear protein which is involved in cell-cycle regulation. The encoded protein is a ubiquitin-ligase capable of ubiquinating PCNP (PEST-containing nuclear protein), and together they may play a role in tumorigenesis. The encoded protein contains an NIRF_N domain, a PHD finger, a set- and ring-associated (SRA) domain, and a RING finger domain and several of these domains have been shown to be essential for the regulation of cell proliferation. This protein may also have a role in intranuclear degradation of polyglutamine aggregates. Alternative splicing results in multiple transcript variants some of which are non-protein coding.
Specifications
| UHRF2 | |
| Polyclonal | |
| Unconjugated | |
| UHRF2 | |
| 2310065A22Rik; AI426270; AW214556; D130071B19Rik; E3 ubiquitin-protein ligase UHRF2; Nirf; np95/ICBP90-like RING finger protein; Np95-like ring finger protein; Nuclear protein 97; nuclear zinc finger protein Np97; RING finger protein 107; RING-type E3 ubiquitin transferase UHRF2; RNF107; RP11-472F14.2; TDRD23; ubiquitin like with PHD and ring finger domains 2; ubiquitin-like PHD and RING finger domain-containing protein 2; ubiquitin-like with PHD and ring finger domains 2, E3 ubiquitin protein ligase; ubiquitin-like, containing PHD and RING finger domains 2; ubiquitin-like, containing PHD and RING finger domains, 2; ubiquitin-like-containing PHD and RING finger domains protein 2; UHRF2; URF2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 109113, 115426, 309331 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| Q7TMI3, Q96PU4 | |
| UHRF2 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human NIRF (15-54aa TIEDVSRKATIEELRERVWALFDVRPECQRLFYRGKQLEN). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction