missing translation for 'onlineSavingsMsg'
Learn More

UHRF1 Antibody (1A1B1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-61838

 View more versions of this product

Product Code. 18202305

  • £435.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

UHRF1 Monoclonal specifically detects UHRF1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

UHRF1
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
E3 ubiquitin-protein ligase UHRF1, EC 6.3.2, EC 6.3.2.-, FLJ21925, hNP95, huNp95, ICBP90NP95, Inverted CCAAT box-binding protein of 90 kDa, Np95, Nuclear protein 95, Nuclear zinc finger protein Np95, RING finger protein 106, RNF106MGC138707, Transcription factor ICBP90, Ubiquitin-like PHD and RING finger domain-containing protein 1, ubiquitin-like with PHD and ring finger domains 1, ubiquitin-like, containing PHD and RING finger domains, 1, Ubiquitin-like-containing PHD and RING finger domains protein 1
Mouse
91.3 kDa
0.1 mL
Primary
Human
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1A1B1
Unconjugated
PBS with 0.05% Sodium Azide
UHRF1
Purified recombinant fragment of human UHRF1 (AA: 616-755) expressed in E. Coli.
Protein G purified
DNA Repair, Zinc Finger
29128
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.