missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UHMK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11624-1-AP-20UL
This item is not returnable.
View return policy
Description
Upon serum stimulation, KIS, a member of the Ser/Thr protein kinase family, phosphorylates CDKN1B/p27Kip1, thus controlling CDKN1B subcellular location and cell cycle progression in G1 phase. This protein, which contains 1 RRM (RNA recognition motif)domain, may be involved in trafficking and/or processing of RNA. KIS is widely expressed, with highest levels in skeletal muscle, kidney, placenta and peripheral blood leukocytes.Specifications
| UHMK1 | |
| Polyclonal | |
| Unconjugated | |
| UHMK1 | |
| KIS, Kist, U2AF homology motif kinase 1, UHMK1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 127933, 16589, 246332 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P97343, Q63285, Q8TAS1 | |
| Uhmk1 | |
| UHMK1 Fusion Protein Ag2204 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction