missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UHMK1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584860
This item is not returnable.
View return policy
Description
Immunogen sequence: NEEEYEDVVE DVKEECQKYG PVVSLLVPKE NPGRGQVFVE YANAGDSKAA QKLLTGRMFD GKFVVATFYP LSAYKRGYLY QTLL Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.
Upon serum stimulation, KIS, a member of the Ser/Thr protein kinase family, phosphorylates CDKN1B/p27Kip1, thus controlling CDKN1B subcellular location and cell cycle progression in G1 phase. This protein, which contains 1 RRM (RNA recognition motif)domain, may be involved in trafficking and/or processing of RNA. KIS is widely expressed, with highest levels in skeletal muscle, kidney, placenta and peripheral blood leukocytes.
Specifications
| UHMK1 | |
| Polyclonal | |
| Unconjugated | |
| UHMK1 | |
| 4732477C12Rik; 4930500M09Rik; AA673513; AI449218; AU021979; kinase interacting stathmin; kinase interacting with leukemia-associated gene (stathmin); kinase interacting with stathmin; KIS; KIS protein kinase; KIST; PAM COOH-terminal interactor protein 2; P-CIP2; Serine/threonine-protein kinase Kist; U2AF homology motif (UHM) kinase 1; U2AF homology motif kinase 1; Uhmk1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 127933 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TAS1 | |
| UHMK1 | |
| Recombinant protein corresponding to Human UHMK1. Recombinant protein control fragment (Product #RP-107800). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction