missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UGT3A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563732
This item is not returnable.
View return policy
Description
Immunogen sequence: IPLFGDQPEN MVRVEAKKFG VSIQLKKLKA ETLALKMKQI M Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 63%.
Specifications
| UGT3A2 | |
| Polyclonal | |
| Unconjugated | |
| UGT3A2 | |
| AI313915; PSEC0073; RGD1564365; UDP glucuronosyltransferase 3 family, polypeptide A2; UDP glycosyltransferase 3 family, polypeptide A2; UDP glycosyltransferase family 3 member A2; UDP glycosyltransferases 3 family, polypeptide A2; UDP-glucuronosyltransferase 3A2; UDPGT 3A2; Ugt3a2; UNQ842/PRO1780 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 167127 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q3SY77 | |
| UGT3A2 | |
| Recombinant protein corresponding to Human UGT3A2. Recombinant protein control fragment (Product #RP-101857). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction