missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGT3A2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93386-0.02ml
This item is not returnable.
View return policy
Description
UGT3A2 Polyclonal antibody specifically detects UGT3A2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| UGT3A2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 2.4.1.17, MGC119426, MGC119429, UDP glycosyltransferase 3 family, polypeptide A2, UDP-glucuronosyltransferase 3A2, UDPGT 3A2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 23-300 of human UGT3A2 (NP_777574.2). AKILTISTVGGSHYLLMDRVSQILQDHGHNVTMLNHKRGPFMPDFKKEEKSYQVISWLAPEDHQREFKKSFDFFLEETLGGRGKFENLLNVLEYLALQCSHFLNRKDIMDSLKNENFDMVIVETFDYCPFLIAEKLGKPFVAILSTSFGSLEFGLPIPLSYVPVFRSLLTDHMDFWGRVKNFLMFFSFCRRQQHMQSTFDNTIKEHFTEGSRPVLSHLLLKAELWFINSDFAFDFARPLLPNTVYVGGLMEKPIKPVPQDLENFIAKFGDSGFVLVTL | |
| 0.02 mL | |
| Lipid and Metabolism | |
| 167127 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction