missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UGT3A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551238
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total UGT3A1 protein.
UDP-glucuronosyltransferases catalyze phase II biotransformation reactions in which lipophilic substrates are conjugated with glucuronic acid to increase water solubility and enhance excretion. They are of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds.
Specifications
| UGT3A1 | |
| Polyclonal | |
| Unconjugated | |
| UGT3A1 | |
| FLJ26528; FLJ34658; UD3A1; UDP glycosyltransferase 3 family, polypeptide A1; UDP glycosyltransferase family 3 member A1; UDP-glucuronosyltransferase 3A1; UDPGT 3A1; UGT3A1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 133688 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q6NUS8 | |
| UGT3A1 | |
| Fusion protein of human UGT3A1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction