missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGT2B7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16661-1-AP-20UL
This item is not returnable.
View return policy
Description
The UGTs (EC 2.4.1.17) serve a major role in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds. UGT2B7 has unique specificity for 3,4-catechol estrogens and estriol, suggesting that it may play an important role in regulating the level and activity of these potent estrogen metabolites. Its subcellular location is the microsome.Specifications
| UGT2B7 | |
| Polyclonal | |
| Unconjugated | |
| UGT2B7 | |
| UDPGT 2B7, UDPGT 2B9, UDPGTh 2, UGT2B7, UGT2B9, UGTB2B9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7364 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P16662 | |
| UGT2B7 | |
| UGT2B7 Fusion Protein Ag10121 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction