missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UGT2B7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576770
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The UGTs serve a major role in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds. UGT2B7 has unique specificity for 3,4-catechol estrogens and estriol, suggesting that it may play an important role in regulating the level and activity of these potent estrogen metabolites. Its subcellular location is the microsome.
Specifications
| UGT2B7 | |
| Polyclonal | |
| Unconjugated | |
| UGT2B7 | |
| 3,4-catechol estrogen specific; 3,4-catechol estrogen-specific UDPGT; UDP glucuronosyltransferase 2 family, polypeptide B7; UDP glucuronosyltransferase 2B7; UDP glucuronosyltransferase family 2 member B7; UDP-glucuronosyltransferase 2B7; UDP-glucuronosyltransferase 2B8; UDP-glucuronosyltransferase 2B9; UDP-glucuronyltransferase, family 2, beta-7; UDPGT; UDPGT 2B7; UDPGT 2B8; UDPGT 2B9; UDPGT2B7; UDPGTh 2; UDPGTH2; UDPGTh-2; Ugt2b7; Ugt2b8; UGT2B9; UGT2B-RH4; UGTB2B9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 286989, 7364 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P16662, Q62789 | |
| UGT2B7 | |
| Recombinant full length Human UGT2B7. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction