missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGT2B7 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00007364-B01P-50ug
This item is not returnable.
View return policy
Description
UGT2B7 Polyclonal antibody specifically detects UGT2B7 in Human samples. It is validated for Western Blot
Specifications
| UGT2B7 | |
| Polyclonal | |
| Western Blot | |
| ABM86740.1 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| UDP glucuronosyltransferase 2 family, polypeptide B73,4-catechol estrogen specific, UDP glycosyltransferase 2 family, polypeptide B7, UGT2B9family 2, beta-7, UGTB2B9 | |
| UGT2B7 (ABM86740.1, 1 a.a. - 529 a.a.) full-length human protein. MSVKWTSVILLIQLSFCFSSGNCGKVLVWAAEYSHWMNIKTILDELIQRGHEVTVLASSASILFDPNNSSALKIEIYPTSLTKTELENFIMQQIKRWSDLPKDTFWLYFSQVQEIMSIFGDITRKFCKDVVSNKKFMKKVQESRFDVIFADAIFPCSELLAELFNIPFVYSLSFSPGYTFEKHSGGFIFPPSYVPVVMSELTDQMTFMERVKNMIYVLYFDFWFEIFDMKKWDQFYSEVLGRPTTLSETMGKADVWLIRNSWNFQFPYPLLPNVDFVGGLHCKPAKPLPKEMEDFVQSSGENGVVVFSLGSMVSNMTEERANVIASALAQIPQKVLWRFDGNKPDTLGLNTRLYKWIPQNDLLGHPKTRAFITHGGANGIYEAIYHGIPMVGIPLFADQPDNIAHMKARGAAVRVDFNTMSSTDLLNALKRVINDPSYKENVMKLSRIQHDQPVKPLDRAVFWIEFVMRHKGAKHLRVAAHDLTWFQYHSLDVIGFLLVCVATVIFIVTKCCLFCFWKFARKAKKGKND | |
| 50 μg | |
| Cancer, Cell Biology, Endocrinology, Signal Transduction | |
| 7364 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction