missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UGT2B4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543384
This item is not returnable.
View return policy
Description
Peptide sequence: NIKTILDELV QRGHEVTVLA SSASISFDPN SPSTLKFEVY PVSLTKTEFE Sequence homology: Human: 100%.
UGT2B4 belongs to the UDP-glycosyltransferase family. UDPGTs are of major importance in the conjugation and subsequent elimination of potentially toxic xenobiotics and endogenous compounds. This isozyme is active on polyhydroxylated estrogens (such as estriol, 4-hydroxyestrone and 2-hydroxyestriol) and xenobiotics (such as 4-methylumbelliferone, 1-naphthol, 4-nitrophenol, 2-aminophenol, 4-hydroxybiphenyl and menthol). It is capable of 6 alpha-hydroxyglucuronidation of hyodeoxycholic acid.
Specifications
| UGT2B4 | |
| Polyclonal | |
| Unconjugated | |
| UGT2B4 | |
| HLUG25; hyodeoxycholic acid; hyodeoxycholic acid-specific UDPGT; UDP glucuronosyltransferase 2 family, polypeptide B15; UDP glucuronosyltransferase 2 family, polypeptide B36; UDP glucuronosyltransferase 2 family, polypeptide B4; UDP glucuronosyltransferase family 2 member B4; UDP glycosyltransferase 2 family, polypeptide B4; UDP-glucuronosyltransferase 2B12; UDP-glucuronosyltransferase 2B15; UDP-glucuronosyltransferase 2B36; UDP-glucuronosyltransferase 2B4; UDP-glucuronyltransferase, family 2, beta-4; UDPGT 2B12; UDPGT 2B15; UDPGT 2B36; UDPGT 2B4; UDPGT2B4; UDPGTH1; UDPGTh-1; UGT2B11; Ugt2b12; Ugt2b15; Ugt2b36; UGT2B4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7363 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P06133 | |
| UGT2B4 | |
| Synthetic peptide directed towards the N-terminal of human UGT2B4 (aa 38-87). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction