missing translation for 'onlineSavingsMsg'
Learn More

UGT2B10 Antibody (2D9), Novus Biologicals™

Product Code. 18361668 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361668 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18361668 Supplier Novus Biologicals Supplier No. H00007365M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UGT2B10 Monoclonal antibody specifically detects UGT2B10 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen UGT2B10
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2D9
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001066
Gene Alias EC 2.4.1.17, MGC142209, UDP glucuronosyltransferase 2 family, polypeptide B10, UDP glucuronosyltransferase 2B10, UDP glycosyltransferase 2 family, polypeptide B10, UDP-glucuronosyltransferase 2B10, UDPGT 2B10, UDPGT2B10
Host Species Mouse
Immunogen UGT2B10 (NP_001066, 62 a.a. ~ 159 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LFDPNDSSTLKLEVYPTSLTKTEFENIIMQLVKRLSEIQKDTFWLPFSQEQEILWAINDIIRNFCKDVVSNKKLMKKLQESRFDIVFADAYLPCGELL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7365
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.