missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGT1A9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69689
This item is not returnable.
View return policy
Description
UGT1A9 Polyclonal specifically detects UGT1A9 in Human samples. It is validated for Western Blot.
Specifications
| UGT1A9 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 2.4.1.17, GNT1, HLUGP4, LUGP4, polypeptide A9, UDP glucuronosyltransferase 1 family, polypeptide A9, UDP-glucuronosyltransferase 1-9, UDP-glucuronosyltransferase 1A9, UDP-glucuronosyltransferase 1-I, UDPGT, UDPGT 1-9, UGT1, UGT1*9, UGT1.9, UGT1-09, UGT1-9, UGT1AI, UGT1I, UGT-1I | |
| Rabbit | |
| 57 kDa | |
| 100 μL | |
| Cancer | |
| 54600 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O60656 | |
| UGT1A9 | |
| Synthetic peptides corresponding to UGT1A9(UDP glucuronosyltransferase 1 family, polypeptide A9) The peptide sequence was selected from the N terminal of UGT1A9. Peptide sequence LLMGSYNDIFDLFFSNCRSLFKDKKLVEYLKESSFDAVFLDPFDNCGLIV. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Sheep: 83%. | |
| Human, Mouse, Rat, Rabbit, Sheep | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction