missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UGT1A6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110546
This item is not returnable.
View return policy
Description
Immunogen sequence: LLLKESKYYT RKIYPVPYDQ EELKNRYQSF GNNHFAERSF LTAPQTEYRN NMIVIGLYFI NCQSLLQDRD TLNFFKESKF DALFTDPALP CGVILAEYLG LPSVYLFRGF PCSLEHTFSR SPDPVSYIPR CYTKFSDHMT FSQRVANFLV NLLEPYLFYC LFSKYEELAS AVLKRDVDII TLYQKVSVWL LRYDFVLEYP RPVMPN.
UGT1A6 is a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This gene is part of a complex locus that encodes several UDP-glucuronosyltransferases. The locus includes thirteen unique alternate first exons followed by four common exons. Four of the alternate first exons are considered pseudogenes. Each of the remaining nine 5' exons may be spliced to the four common exons, resulting in nine proteins with different N-termini and identical C-termini. Each first exon encodes the substrate binding site, and is regulated by its own promoter. The enzyme is active on phenolic and planar compounds. Alternative splicing in the unique 5' end of this gene results in two transcript variants.
Specifications
| UGT1A6 | |
| Polyclonal | |
| Unconjugated | |
| UGT1A6 | |
| A1; GNT1; HLUGP; HLUGP1; MGC29860; Phenol UDP-glucuronosyltransferase; phenol-metabolizing UDP-glucuronosyltransferase; P-nitrophenol specific; P-nitrophenol-specific UDPGT; UDP glucuronosyltransferase 1 family, polypeptide A6; UDP glucuronosyltransferase 1 family, polypeptide A6A; UDP glucuronosyltransferase 1A6; UDP glucuronosyltransferase family 1 member A6; UDP glycosyltransferase 1 family polypeptide A4; UDP glycosyltransferase 1 family, polypeptide A6; UDP-glucuronosyltransferase; UDP-glucuronosyltransferase 1 family polypeptide A6s; UDP-glucuronosyltransferase 1-6; UDP-glucuronosyltransferase 1A6; UDP-glucuronosyltransferase 1-F; Udpgt; UDPGT 1-6; UGP1A1; UGT1; UGT1*6; UGT1.6; UGT1-06; Ugt1a6; Ugt1a6a; UGT1A6S; UGT1A7; UGT1F; UGT-1F | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 54578, 94284 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P19224, Q64435 | |
| UGT1A6, Ugt1a6a | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 65-270 of human UGT1A6 (NP_001063.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction