missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGT1A6 Polyclonal Antibody

Brand: Thermo Scientific PA5116104
This item is not returnable.
View return policy
Description
Specifications
| UGT1A6 | |
| Polyclonal | |
| Unconjugated | |
| UGT1A6 | |
| A1; GNT1; HLUGP; HLUGP1; MGC29860; Phenol UDP-glucuronosyltransferase; phenol-metabolizing UDP-glucuronosyltransferase; P-nitrophenol specific; P-nitrophenol-specific UDPGT; UDP glucuronosyltransferase 1 family, polypeptide A6; UDP glucuronosyltransferase 1 family, polypeptide A6A; UDP glucuronosyltransferase 1A6; UDP glucuronosyltransferase family 1 member A6; UDP glycosyltransferase 1 family polypeptide A4; UDP glycosyltransferase 1 family, polypeptide A6; UDP-glucuronosyltransferase; UDP-glucuronosyltransferase 1 family polypeptide A6s; UDP-glucuronosyltransferase 1-6; UDP-glucuronosyltransferase 1A6; UDP-glucuronosyltransferase 1-F; Udpgt; UDPGT 1-6; UGP1A1; UGT1; UGT1*6; UGT1.6; UGT1-06; Ugt1a6; Ugt1a6a; UGT1A6S; UGT1A7; UGT1F; UGT-1F | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 113992, 54578, 94284 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P08430, P19224, Q64435 | |
| UGT1A6, Ugt1a6a | |
| A synthesized peptide derived from human UGT1A6(Accession P19224), corresponding to amino acid residues V60-T110. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction