missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UGT1A5 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA543353

Product Code. 15526012

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Peptide sequence: EENFFTLTTY AISWTQDEFD RLLLGHTQSF FETEHLLMKF SRRMAIMNNM Sequence homology: Human: 100%; Rat: 83%.

This gene encodes a UDP-glucuronosyltransferase, an enzyme of the glucuronidation pathway that transforms small lipophilic molecules, such as steroids, bilirubin, hormones, and drugs, into water-soluble, excretable metabolites. This gene is part of a complex locus that encodes several UDP-glucuronosyltransferases. The locus includes thirteen unique alternate first exons followed by four common exons. Four of the alternate first exons are considered pseudogenes. Each of the remaining nine 5' exons may be spliced to the four common exons, resulting in nine proteins with different N-termini and identical C-termini. Each first exon encodes the substrate binding site, and is regulated by its own promoter.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

UGT1A5
Polyclonal
Unconjugated
UGT1A5
B5; GNT1; UDP glucuronosyltransferase 1 family, polypeptide A5; UDP glucuronosyltransferase family 1 member A5; UDP glycosyltransferase 1 family, polypeptide A5; UDP-glucuronosyltransferase 1 family polypeptide A5s; UDP-glucuronosyltransferase 1-5; UDP-glucuronosyltransferase 1A5; UDP-glucuronosyltransferase 1-E; UDPGT; UDPGT 1-5; Ugt1; UGT1*5; UGT1.5; UGT1-05; UGT1A5; Ugt1a6; UGT1E; UGT-1E
Rabbit
Affinity Chromatography
RUO
54579
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P35504
UGT1A5
Synthetic peptide directed towards the N-terminal of human UGT1A5 (aa 71-120).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.