missing translation for 'onlineSavingsMsg'
Learn More

UGT1A3 Antibody (1C10), Novus Biologicals™

Product Code. 18328459 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328459 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328459 Supplier Novus Biologicals Supplier No. H00054659M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

UGT1A3 Monoclonal antibody specifically detects UGT1A3 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen UGT1A3
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1C10
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_061966
Gene Alias GNT1, UDP glucuronosyltransferase 1 family, polypeptide A3, UDP glucuronosyltransferase 1A3, UDP glycosyltransferase 1 family, polypeptide A3, UDP-glucuronosyltransferase 1-3, UDP-glucuronosyltransferase 1A3, UDP-glucuronosyltransferase 1-C, UDPGT, UDPGT 1-3, UGT1, UGT1*3, UGT1.3, UGT1-03, UGT-1C, UGT1CEC 2.4.1.17
Host Species Mouse
Immunogen UGT1A3 (NP_061966, 30 a.a. ~ 129 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KVLVVPIDGSHWLSMREVLRELHARGHQAVVLTPEVNMHIKEENFFTLTTYAISWTQDEFDRHVLGHTQLYFETEHFLKKFFRSMAMLNNMSLVYHRSCV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 54659
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.