missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGT Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £375.00
Specifications
| Antigen | UGT |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18693431
|
Novus Biologicals
NBP2-93330-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18687541
|
Novus Biologicals
NBP2-93330-0.1ml |
0.1 mL |
£375.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UGT Polyclonal antibody specifically detects UGT in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| UGT | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| solute carrier family 35 (UDP-galactose transporter), member 2, solute carrier family 35 (UDP-galactose transporter), member A2, UDP-galactose translocator, UGALTUGAT, UGT1, UGT2UDP-galactose transporter, UGTLSolute carrier family 35 member A2, UGTUDP-Gal-Tr | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-242 of human SLC35A2 (NP_001027460.1). MAAVGAGGSTAAPGPGAVSAGALEPGTASAAHRRLKYISLAVLVVQNASLILSIRYARTLPGDRFFATTAVVMAEVLKGLTCLLLLFAQKRGNVKHLVLFLHEAVLVQYVDTLKLAVPSLIYTLQNNLQYVAISNLPAATFQPSPRCSQSHSLCLCLRLRALRSPAASRAATTTAAVFPPWRPHHGALSAKVSAGEVRAGSNGGTQGRGTGVEGVGHLQDPSRHPPGPGSSGFGRWSFLPGH | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 7355 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title