missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGT Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98137AF532
This item is not returnable.
View return policy
Description
UGT Polyclonal antibody specifically detects UGT in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| UGT | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 7355 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| solute carrier family 35 (UDP-galactose transporter), member 2, solute carrier family 35 (UDP-galactose transporter), member A2, UDP-galactose translocator, UGALTUGAT, UGT1, UGT2UDP-galactose transporter, UGTLSolute carrier family 35 member A2, UGTUDP-Gal-Tr | |
| Produced in rabbits immunized with E. coli-derived Human UGT fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction