missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGCG Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £364.00
Specifications
| Antigen | UGCG |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
UGCG Polyclonal antibody specifically detects UGCG in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| UGCG | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Endocrinology | |
| PBS (pH 7.3), 50% glycerol | |
| 7357 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ceramide glucosyltransferase, EC 2.4.1.80, GCSGLCT1, GLCT-1, Glucosylceramide synthase, UDP-glucose ceramide glucosyltransferaseUDP-glucose:N-acylsphingosine D-glucosyltransferase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 33-195 of human UGCG (NP_003349.1). TRLHLNKKATDKQPYSKLPGVSLLKPLKGVDPNLINNLETFFELDYPKYEVLLCVQDHDDPAIDVCKKLLGKYPNVDARLFIGGKKVGINPKINNLMPGYEVAKYDLIWICDSGIRVIPDTLTDMVNQMTEKVGLVHGLPYVADRQGFAATLEQVYFGTSHPR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title