missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UFM1 Recombinant Rabbit Monoclonal Antibody (K01_1K41)

Product Code. 30826945
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826945 100 μL
30825950 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826945 Supplier Invitrogen™ Supplier No. MA563152

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

UFM1 Recombinant Monoclonal Antibody for IHC (P), Western Blot

UFM1 is a ubiquitin-like protein that is conjugated to target proteins by E1-like activating enzyme UBA5 (UBE1DC1; MIM 610552) and E2-like conjugating enzyme UFC1 (MIM 610554) in a manner analogous to ubiquitylation (see UBE2M; MIM 603173) (Komatsu et al, 2004).
TRUSTED_SUSTAINABILITY

Specifications

Antigen UFM1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1K41
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene Ufm1
Gene Accession No. P61960, P61961
Gene Alias 1810045K17Rik; AI132708; AI463323; bA131P10.1; BM-002; C13orf20; ENSMUSG00000074598; Gm10726; hypothetical protein BM-002; RGD1304890; similar to ubiquitin-fold modifier 1; ubfm1; ubfm1 protein; ubiquitin fold modifier 1; ubiquitin fold modifier 1; ubiquitin-fold modifier 1; ubiquitin-fold modifier 1; ufm1; wu:fa10f05; wu:fb72g01; zBM-002; zgc:55335
Gene Symbols Ufm1
Host Species Rabbit
Immunogen A synthetic peptide of human UFM1.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51569, 67890
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.