missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UFM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558916
This item is not returnable.
View return policy
Description
Immunogen sequence: IRAFPTTTPR SLHLFTSSTF LARALPGAFP TGACEERLSV PESTPFTAVL KFAAEEFKVP AATSAIITND GIGINPAQTA GNVFLKHGSE LRIIPRDRVG SC Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 63%.
UFM1 is a ubiquitin-like protein that is conjugated to target proteins by E1-like activating enzyme UBA5 (UBE1DC1; MIM 610552) and E2-like conjugating enzyme UFC1 (MIM 610554) in a manner analogous to ubiquitylation (see UBE2M; MIM 603173) (Komatsu et al, 2004).
Specifications
| UFM1 | |
| Polyclonal | |
| Unconjugated | |
| Ufm1 | |
| 1810045K17Rik; AI132708; AI463323; bA131P10.1; BM-002; C13orf20; ENSMUSG00000074598; Gm10726; hypothetical protein BM-002; RGD1304890; similar to ubiquitin-fold modifier 1; ubfm1; ubfm1 protein; ubiquitin fold modifier 1; ubiquitin fold modifier 1; ubiquitin-fold modifier 1; ubiquitin-fold modifier 1; ufm1; wu:fa10f05; wu:fb72g01; zBM-002; zgc:55335 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51569 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P61960 | |
| Ufm1 | |
| Recombinant protein corresponding to Human UFM1. Recombinant protein control fragment (Product #RP-97931). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction