missing translation for 'onlineSavingsMsg'
Learn More

UFM1 Antibody [HRP], Novus Biologicals™

Product Code. 30734421 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734421 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734421 Supplier Novus Biologicals Supplier No. AF8237H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

UFM1 Polyclonal antibody specifically detects UFM1 in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen UFM1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias bA131P10.1, BM-002, C13orf20, chromosome 13 open reading frame 20, ubiquitin-fold modifier 1
Host Species Sheep
Immunogen E. coli-derived recombinant human UFM1, Met1-Gly83, Accession # P61960
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 51569
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.