missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UFM1 Activating Enzyme/UBA5 Polyclonal antibody specifically detects UFM1 Activating Enzyme/UBA5 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | UFM1 Activating Enzyme/UBA5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 |
| Formulation | PBS (pH 7.4) |
| Gene Accession No. | NP_079094.1 |
| Gene Alias | FLJ17281, FLJ23251UBA5, ThiFP1, UBE1DC1ubiquitin-activating enzyme E1 homolog, ubiquitin-like modifier activating enzyme 5, ubiquitin-like modifier-activating enzyme 5, UFM1-activating enzyme |
| Host Species | Mouse |
| Immunogen | UBE1DC1 (NP_079094.1, 1 a.a. - 404 a.a.) full-length human protein. MAESVERLQQRVQELERELAQERSLQVPRSGDGGGGRVRIEKMSSEVVDSNPYSRLMALKRMGIVSDYEKIRTFAVAIVGVGGVGSVTAEMLTRCGIGKLLLFDYDKVELANMNRLFFQPHQAGLSKVQAAEHTLRNINPDVLFEVHNYNITTVENFQHFMDRISNGGLEEGKPVDLVLSCVDNFEARMTINTACNELGQTWMESGVSENAVSGHIQLIIPGESACFACAPPLVVAANIDEKTLKREGVCAASLPTTMGVVAGILVQNVLKFLLNFGTVSFYLGYNAMQDFFPTMSMKPNPQCDDRNCRKQQEEYKKKVAALPKQEVIQEEEEIIHEDNEWGIELVSEVSEEELKNFSGPVPDLPEGITVAYTIPKKQEDSVTELTVEDSGESLEDLMAKMKNM |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?