missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UFM1 Activating Enzyme/UBA5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | UFM1 Activating Enzyme/UBA5 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
UFM1 Activating Enzyme/UBA5 Polyclonal specifically detects UFM1 Activating Enzyme/UBA5 in Human samples. It is validated for Western Blot.Specifications
| UFM1 Activating Enzyme/UBA5 | |
| Polyclonal | |
| Rabbit | |
| Q9GZZ9 | |
| 79876 | |
| Synthetic peptides corresponding to UBA5(ubiquitin-like modifier activating enzyme 5) The peptide sequence was selected from the middle region of UBA5. Peptide sequence VLSCVDNFEARMTINTACNELGQTWMESGVSENAVSGHIQLIIPGESACF. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| FLJ17281, FLJ23251UBA5, ThiFP1, UBE1DC1ubiquitin-activating enzyme E1 homolog, ubiquitin-like modifier activating enzyme 5, ubiquitin-like modifier-activating enzyme 5, UFM1-activating enzyme | |
| UBA5 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title