missing translation for 'onlineSavingsMsg'
Learn More

Ufenamate, MedChemExpress

Product Code. 16474196
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474196 100 mg 100mg
16445477 5 mg 5mg
16429436 50 mg 50mg
16422738 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16474196 Supplier Medchem Express Supplier No. HY100009
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ufenamate (Flufenamic acid butyl ester) is an anthranilic acid-based anti-inflammatory agent. Ufenamate can be used for the research of skin diseases, such as acute and chronic eczema, contact dermatitis, diaper dermatitis, miliaria and atopic dermatitis.

Ufenamate (Flufenamic acid butyl ester) is an anthranilic acid-based anti-inflammatory agent. Ufenamate can be used for the research of skin diseases, such as acute and chronic eczema, contact dermatitis, diaper dermatitis, miliaria and atopic dermatitis.

Specifications

Chemical Name or Material Ufenamate
Physical Form Liquid
CAS 67330-25-0
Color Colorless
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H18F3NO2
For Use With (Application) COVID-19-immunoregulation
Synonym Flufenamic acid butyl ester Butyl flufenamate
Solubility Information DMSO : ≥ 44 mg/mL (130.43 mM)
SMILES O=C(OCCCC)C1=CC=CC=C1NC2=CC=CC(C(F)(F)F)=C2
Molecular Weight (g/mol) 337.34
Formula Weight 337.34
Percent Purity 99.85%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.