missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UFD1L Antibody (2A6F3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £441.00
Specifications
| Antigen | UFD1L |
|---|---|
| Clone | 2A6F3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18271531
|
Novus Biologicals
NBP2-61829 |
0.1 mL |
£441.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18641118
|
Novus Biologicals
NBP2-61829-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UFD1L Monoclonal specifically detects UFD1L in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| UFD1L | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| 7353 | |
| Purified recombinant fragment of human UFD1L (AA: 208-307) expressed in E. Coli. | |
| Primary | |
| 34.5 kDa |
| 2A6F3 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| UFD1L | |
| IgG2b | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title