missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UFC1 Recombinant Rabbit Monoclonal Antibody (23GB4245), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553716
This item is not returnable.
View return policy
Description
UFC1 is an E2-like conjugating enzyme for ubiquitin-fold modifier-1 (Komatsu et al., 2004 ).
Specifications
| UFC1 | |
| Recombinant Monoclonal | |
| 2.0 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9Y3C8 | |
| UFC1 | |
| A synthesized peptide derived from UFC1 (1-130AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4245 | |
| Unconjugated | |
| UFC1 | |
| 1110021H02Rik; AA062310; CGI-126; ESTM29; HSPC155; ubiquitin-fold modifier conjugating enzyme 1; ubiquitin-fold modifier-conjugating enzyme 1; UFC1; ufc1-s; Ufm1-conjugating enzyme 1; Ufm1-conjugating enzyme 1- | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 51506 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction