missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UFC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15783-1-AP-150UL
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Product No. 15783-1-AP specifically recognizes the 21 kDa protein.
UFC1 is an E2-like conjugating enzyme for ubiquitin-fold modifier-1 (Komatsu et al., 2004 ).Tekniske data
| UFC1 | |
| Polyclonal | |
| Unconjugated | |
| UFC1 | |
| HSPC155, UFC1, Ufm1 conjugating enzyme 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 445268, 51506, 66155 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6BBI8, Q9CR09, Q9Y3C8 | |
| UFC1 | |
| UFC1 Fusion Protein Ag8480 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion