missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UFC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15783-1-AP-20UL
This item is not returnable.
View return policy
Description
Product No. 15783-1-AP specifically recognizes the 21 kDa protein.
UFC1 is an E2-like conjugating enzyme for ubiquitin-fold modifier-1 (Komatsu et al., 2004 ).Specifications
| UFC1 | |
| Polyclonal | |
| Unconjugated | |
| UFC1 | |
| HSPC155, UFC1, Ufm1 conjugating enzyme 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 445268, 51506, 66155 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6BBI8, Q9CR09, Q9Y3C8 | |
| UFC1 | |
| UFC1 Fusion Protein Ag8480 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction