missing translation for 'onlineSavingsMsg'
Learn More

UF010, MedChemExpress

Product Code. 16497566
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497566 100 mg 100mg
16477668 10 mg 10mg
16473677 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497566 Supplier Medchem Express Supplier No. HY18976
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress UF010 is a potent and selective HDAC inhibitor with IC50 ∼0.06 μM, 0.1 μM, 0.5 μM and 1.5 μM for HDACs 3, 2, 1 and 8, respectively.

UF010 is a potent and selective HDAC inhibitor with IC50 ∼0.06 μM, 0.1 μM, 0.5 μM and 1.5 μM for HDACs 3, 2, 1 and 8, respectively.

Specifications

Chemical Name or Material UF010
CAS 537672-41-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H15BrN2O
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (368.80 mM)
SMILES O=C(NNCCCC)C1=CC=C(Br)C=C1
Molecular Weight (g/mol) 271.15
Formula Weight 271.15
Percent Purity 99.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.