missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UEVLD Polyclonal antibody specifically detects UEVLD in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | UEVLD |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Gene Alias | Attp, EV and lactate/malate dehydrogenase domain-containing protein, signaling molecule ATTP, ubiquitin-conjugating enzyme E2 variant 3, UEV and lactate/malate dehyrogenase domains, UEV-3, UEV3FLJ11068 |
| Gene ID (Entrez) | 55293 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?