missing translation for 'onlineSavingsMsg'
Learn More

UEV2 Antibody [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30763878 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30763878 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30763878 Supplier Novus Biologicals Supplier No. NBP347078MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. Se returpolicy

Rabbit Polyclonal Antibody

UEV2 Polyclonal antibody specifically detects UEV2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen UEV2
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 1 alpha, 25-dihydroxyvitamin D3-inducible, DDVit-1, DDVIT1, EDAF-1, EDPF1, EDPF-1DDVit 1, enterocyte differentiation promoting factor, Enterocyte differentiation-associated factor 1, enterocyte differentiation-associated factor EDAF-1, Enterocyte differentiation-promoting factor 1, methyl methanesulfonate sensitive 2, S. cerevisiae, homolog of, MMS2MMS2 homolog, ubiquitin-conjugating enzyme E2 variant 2, UEV2, UEV-2, Vitamin D3-inducible protein
Host Species Rabbit
Immunogen ubiquitin-conjugating enzyme E2 variant 2. (Uniprot# Q15819)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7336
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.