missing translation for 'onlineSavingsMsg'
Learn More

UDP glucose dehydrogenase Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18397802 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397802 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397802 Supplier Novus Biologicals Supplier No. NBP306479AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UDP glucose dehydrogenase Polyclonal antibody specifically detects UDP glucose dehydrogenase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen UDP glucose dehydrogenase
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias EC 1.1.1.22, GDH, UDPGDHUDP-glucose dehydrogenase, UDP-Glc dehydrogenase, UDP-GlcDH, UDP-glucose 6-dehydrogenase, UGD, uridine diphospho-glucose dehydrogenase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human UDP glucose dehydrogenase fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7358
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.