missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UCRC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588641
This item is not returnable.
View return policy
Description
Immunogen sequence: MAAATLTSKL YSLLFRRTST FALTIIVGVM FFERAFDQGA DAIYDHINEG KLWKHIKHKY ENK; Positive Samples: 22RV1, H460, Mouse kidney, Rat heart; Cellular Location: Mitochondrion inner membrane.
UCRC is a subunit of mitochondrial complex III (ubiquinol-cytochrome c reductase; EC 1.10.2.2), which forms the middle segment of the respiratory chain of the inner mitochondrial membrane. UCRC is a subunit of mitochondrial complex III (ubiquinol-cytochrome c reductase; EC 1.10.2.2), which forms the middle segment of the respiratory chain of the inner mitochondrial membrane (Schagger et al., 1995 ).
Specifications
| UCRC | |
| Polyclonal | |
| Unconjugated | |
| UQCR10 | |
| 1110020P15Rik; AA960494; Complex III subunit 9; complex III subunit X; Cytochrome b-c1 complex subunit 9; cytochrome c1 non-heme 7 kDa protein; cytochrome C1, nonheme 7kDa protein; HSPC051; HSPC119; HSPC151; QCR9; ubiquinol-cytochrome c reductase complex (7.2 kD); ubiquinol-cytochrome c reductase complex 7.2 kDa protein; ubiquinol-cytochrome c reductase complex 7.2kDa protein; ubiquinol-cytochrome c reductase complex protein; ubiquinol-cytochrome c reductase, complex III subunit X; ubiquinol-cytochrome c reductase, complex III subunit X, 7.2kDa; UCCR7.2; Ucrc; Unknown (protein for MGC:127828); UQCR10 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 29796, 66152, 685322 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8R1I1, Q9UDW1 | |
| UQCR10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-63 of human UQCR10 (NP_0375192). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction