missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCP4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £375.00
Specifications
| Antigen | UCP4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18667290
|
Novus Biologicals
NBP2-94703-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18685621
|
Novus Biologicals
NBP2-94703-0.1ml |
0.1 mL |
£375.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UCP4 Polyclonal antibody specifically detects UCP4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| UCP4 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 9481 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ33552, mitochondrial uncoupling protein 4, Solute carrier family 25 member 27, solute carrier family 25, member 27, UCP 4, UCP4uncoupling protein 4 | |
| A synthetic peptide corresponding to a sequence within amino acids 201-300 of human UCP4 (NP_004268.3). NQPRDKQGRGLLYKSSTD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title