missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UCP3 Recombinant Rabbit Monoclonal Antibody (10I0J9)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555374
This item is not returnable.
View return policy
Description
Immunogen Sequence: YDILKEKLLD YHLLTDNFPC HFVSAFGAGF CATVVASPVD VVKTRYMNSP PGQYFSPLDC MIKMVAQEGP TAFYKGFTPS FLRLGSWNVV MFVTYEQLKR A.
The uncoupling protein UCP1 (formerly designated UCP) is an integral membrane protein unique to brown adipose tissue mitochondria. UCP1 forms a dimer that acts as a proton channel, which can uncouple oxidative phosphorylation by dissipating the electrochemical potential across the inner mitochondrial membrane. This process induces heat production in brown adipose tissue and is involved in regulation of body temperature and glucose metabolism. UCP2 is a structurally related protein that also uncouples mitochondrial respiration. It is more widely expressed in human and mouse tissues, including white adipose tissue and muscle, than is UCP1. UCP2 is thought to play a role in body weight regulation. An additional UCP family member, UCP3, is highly muscle specific and is possibly involved in the uncoupling of oxidative phosphorylation in skeletal muscle.
Specifications
| UCP3 | |
| Recombinant Monoclonal | |
| 1.25 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P56501 | |
| Ucp3 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 10I0J9 | |
| Unconjugated | |
| Ucp3 | |
| AI645527; mitochondrial uncoupling protein 3; Slc25a9; solute carrier family 25 member 9; UCP 3; UCP3; UCP-3; uncoupling protein; uncoupling protein 3; uncoupling protein 3 (mitochondrial, proton carrier); uncoupling protein 3, mitochondrial | |
| Rabbit | |
| Protein A | |
| RUO | |
| 22229 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction