missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577553
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20C upon receipt. Reconstitute with 50 μL deionized water.
UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. UCP2 gene is expressed in many tissues, with the greatest expression in skeletal muscle. UCP2 is thought to play a role in non shivering thermogenesis, obesity and diabetes.
Specifications
| UCP2 | |
| Polyclonal | |
| Unconjugated | |
| UCP2 | |
| Mitochondrial Uncoupling Protein 2 (UCP2) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22228, 54315, 7351 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P55851, P56500, P70406 | |
| Ucp2 | |
| Peptide (C)GRRYQSTVEAYK, corresponding to amino acid residues 153-164 of rat mitochondrial uncoupling protein 2 (UCP2) | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction