missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UCP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580203
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat spleen tissue, rat cardiac muscle tissue, rat brain tissue, mouse spleen tissue, mouse cardiac muscle tissue, mouse brain tissue, human Hela whole cell.
UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. UCP2 gene is expressed in many tissues, with the greatest expression in skeletal muscle. UCP2 is thought to play a role in non shivering thermogenesis, obesity and diabetes.
Specifications
| UCP2 | |
| Polyclonal | |
| Unconjugated | |
| Ucp2 | |
| BMIQ4; Mitochondrial uncoupling protein 2; Slc25a8; Solute carrier family 25 member 8; UCP 2; Ucp2; UCPH; uncoupling protein; uncoupling protein 2; uncoupling protein 2 (mitochondrial, proton carrier); uncoupling protein 2, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22228, 54315, 7351 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P55851, P56500, P70406 | |
| Ucp2 | |
| A synthetic peptide corresponding to a sequence in the middle region of human UCP2 (134-170aa AQPTDVVKVRFQAQARAGGGRRYQSTVNAYKTIAREE). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction