missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UCP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16699624
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16699624 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16699624 Supplier Invitrogen™ Supplier No. PA5103176

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total UCP2.

UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. UCP2 gene is expressed in many tissues, with the greatest expression in skeletal muscle. UCP2 is thought to play a role in non shivering thermogenesis, obesity and diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ucp2
Gene Accession No. P55851, P56500, P70406
Gene Alias BMIQ4; Mitochondrial uncoupling protein 2; Slc25a8; Solute carrier family 25 member 8; UCP 2; Ucp2; UCPH; uncoupling protein; uncoupling protein 2; uncoupling protein 2 (mitochondrial, proton carrier); uncoupling protein 2, mitochondrial
Gene Symbols Ucp2
Host Species Rabbit
Immunogen A synthesized peptide derived from human UCP2(Accession P55851), corresponding to amino acid residues N190-T240.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22228, 54315, 7351
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.