missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UCP2 Monoclonal Antibody (D7), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543550
This item is not returnable.
View return policy
Description
UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. UCP2 gene is expressed in many tissues, with the greatest expression in skeletal muscle. UCP2 is thought to play a role in non shivering thermogenesis, obesity and diabetes.
Specifications
| UCP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P55851 | |
| Ucp2 | |
| Arg154-Val289 (Accession P55851), with N-terminal His Tag. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b λ |
| Immunohistochemistry (Paraffin), Western Blot | |
| D7 | |
| Unconjugated | |
| Ucp2 | |
| BMIQ4; Mitochondrial uncoupling protein 2; Slc25a8; Solute carrier family 25 member 8; UCP 2; Ucp2; UCPH; uncoupling protein; uncoupling protein 2; uncoupling protein 2 (mitochondrial, proton carrier); uncoupling protein 2, mitochondrial | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 7351 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction