missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCP2 Antibody (6C8B3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52581-0.025mg
This item is not returnable.
View return policy
Description
UCP2 Monoclonal specifically detects UCP2 in Human samples. It is validated for Western Blot, ELISA, CyTOF-ready.
Specifications
| UCP2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, CyTOF-ready | |
| mitochondrial uncoupling protein 2, SLC25A8BMIQ4, Solute carrier family 25 member 8, UCP 2, UCPH, uncoupling protein 2 (mitochondrial, proton carrier) | |
| Mouse | |
| 33.2 kDa | |
| 0.025 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, CyTOF, Immunohistochemistry, Immunocytochemistry | |
| 6C8B3 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| UCP2 | |
| Purified recombinant fragment of human UCP2 (AA: 1-309) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| 7351 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction