missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCP2 Antibody (3F1B9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £438.00
Specifications
| Antigen | UCP2 |
|---|---|
| Clone | 3F1B9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18167899
|
Novus Biologicals
NBP2-52582 |
0.1 mg |
£438.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623879
|
Novus Biologicals
NBP2-52582-0.025mg |
0.025 mg |
£173.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UCP2 Monoclonal specifically detects UCP2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
| UCP2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| mitochondrial uncoupling protein 2, SLC25A8BMIQ4, Solute carrier family 25 member 8, UCP 2, UCPH, uncoupling protein 2 (mitochondrial, proton carrier) | |
| UCP2 | |
| IgG2 α | |
| Protein A or G purified |
| 3F1B9 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 7351 | |
| Purified recombinant fragment of human UCP2 (AA: 1-309) expressed in E. Coli. | |
| Primary | |
| 33.2 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title