missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UCP1 Recombinant Rabbit Monoclonal Antibody (HL2944)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556908
This item is not returnable.
View return policy
Description
UCP1 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Mitochondrial uncoupling proteins are members of the family of mitochondrial anion carrier proteins. UCPs separate oxidative phosphorylation from ATP synthesis with energy dissipated as heat, also referred to as the mitochondrial proton leak. UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. Tissue specificity occurs for the different UCPs and the exact methods of how UCPs transfer H+/OH- are not known. UCPs contain the three homologous protein domains of MACPs. This gene is expressed only in brown adipose tissue, a specialized tissue which functions to produce heat.
Specifications
| UCP1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P12242 | |
| Ucp1 | |
| Recombinant fragment of human UCP1. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2944 | |
| Unconjugated | |
| Ucp1 | |
| AI385626; HGNC:12517; mitochondrial brown fat uncoupling protein 1; SLC25A7; solute carrier family 25 member 7; Thermogenin; Ucp; UCP 1; Ucp1; Ucpa; uncoupling protein; uncoupling protein 1; uncoupling protein 1 (mitochondrial, proton carrier); uncoupling protein 1, mitochondrial; uncoupling protein, mitochondrial; Uncp; unnamed protein product | |
| Rabbit | |
| Protein A | |
| RUO | |
| 22227 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction