missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-20796
This item is not returnable.
View return policy
Description
UCP1 Polyclonal specifically detects UCP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications
| UCP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS pH7, 1% BSA, 20% Glycerol with 0.025% Proclin 300 | |
| SLC25A7mitochondrial brown fat uncoupling protein 1, Solute carrier family 25 member 7, thermogenin, UCP, UCP 1, uncoupling protein 1 (mitochondrial, proton carrier) | |
| Rabbit | |
| 33 kDa | |
| 0.1 mL | |
| Cancer, Cellular Signaling, metabolism | |
| 7350 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.28mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P25874 | |
| UCP1 | |
| Recombinant protein encompassing a sequence within the center region of human UCP1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction