missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UCMA Polyclonal Antibody
Description
A suggested positive control is SW1353 cell lysate. PA5-20768 can be used with blocking peptide PEP-0882.
UCMA is a gene that encodes for a protein that is specific to chondrocytes and highly charged. This protein is expressed abundantly in the upper immature zone of fetal and juvenile epiphyseal cartilage. The encoded protein undergoes proteolytic processing to generate a mature protein that is secreted into the extracellular matrix. The glutamic acid residues in the protein undergo gamma carboxylation in a vitamin K-dependent manner. Undercarboxylation of the protein has been associated with osteoarthritis in humans. Alternative splicing of the gene results in multiple transcript variants that encode different isoforms. UCMA is also associated with diseases such as Osteoarthritis and Poland Syndrome.
Specifications
Specifications
| Antigen | UCMA |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | UCMA |
| Gene Accession No. | Q14BU0, Q8WVF2 |
| Gene Alias | 1110017I16Rik; AW121955; C10orf49; Gla-rich protein; GRP; GRP/UCMA; RGD1308977; Ucma; ucma {ECO:0000250; Ucma-C; UniProtKB:Q14BU0}; Unique cartilage matrix-associated protein; unique cartilage matrix-associated protein {ECO:0000250; Unique cartilage matrix-associated protein C-terminal fragment; upper zone of growth plate and cartilage matrix associated; upper zone of growth plate and cartilage matrix associated protein |
| Gene Symbols | UCMA |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?