missing translation for 'onlineSavingsMsg'
Learn More

UCMA Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18409926 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409926 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409926 Supplier Novus Biologicals Supplier No. NBP176321MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

UCMA Polyclonal antibody specifically detects UCMA in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen UCMA
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias C10orf49, chromosome 10 open reading frame 49, Gla-rich protein, GRP, unique cartilage matrix-associated protein, upper zone of growth plate and cartilage matrix associated
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human UCMA. The immunogen is located within the last 50 amino acids of UCMA. Amino Acid Squence: WHYDGLHPSYLYNRHHT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221044
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.