missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ UCLA GP130 2

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
UCLA GP130 2 is a gp130 agonist. Protects against NMDA-induced toxicity in primary hippocampal neurons in vitro. Increases STAT3 phosphorylation in SH-SY5Y cells. Brain penetrant.
Specifications
Specifications
| Chemical Name or Material | N-(4-Fluorophenyl)-4-phenyl-2-thiazolamine |
| CAS | 339303-87-6 |
| Quantity | 50 mg |
| Molecular Formula | C15H11FN2S |
| Formula Weight | 270.32 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction